Learn More
Medchemexpress LLC Panasenoside | 31512-06-8 | 1 MG
SDP
Catalog No. 5000420577
Encompass_Preferred
Panasenoside is a flavonoid isolated from *Lilium pumilum* DC. It exhibits α-glucosidase inhibitory activity, making it suitable for research applications. It is provided as a solid with high purity.
  • Exhibits α-glucosidase inhibitory activity
  • Isolated from *Lilium pumilum* DC
  • High purity solid appearance
  • For research use only
Catalog No. 50-004-20577 Supplier Medchemexpress LLC Supplier No. HYN42581MG
Panasenoside is a flavonoid isolated from *Lilium pumilum* DC. It exhibits α-glucosidase inhibitory activity, making it suitable for research applications. It is provided as a solid with high purity.
  • Exhibits α-glucosidase inhibitory activity
  • Isolated from *Lilium pumilum* DC
  • High purity solid appearance
  • For research use only