Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Panasenoside | 31512-06-8 | MFCD30725484 | 5 MG
SDP

Catalog No. 5000420919
Encompass_Preferred

Panasenoside is a flavonoid isolated from *Lilium pumilum* DC. and *Panax ginseng* C. A. Meyer. This white to yellow solid exhibits α-glucosidase inhibitory activity. It is recommended to store this product at 4°C, protected from light. When in solvent, store at -80°C for up to 6 months, or at -20°C for up to 1 month, also protected from light.

  • Inhibits α-glucosidase
  • Isolated from natural plant sources
  • High purity of 99.79%
  • Appearance: White to yellow solid
  • Molecular weight: 610.52
  • Chemical formula: C27H30O16
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-20919 Supplier Medchemexpress LLC Supplier No. HYN42585MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Panasenoside is a flavonoid isolated from *Lilium pumilum* DC. and *Panax ginseng* C. A. Meyer. This white to yellow solid exhibits α-glucosidase inhibitory activity. It is recommended to store this product at 4°C, protected from light. When in solvent, store at -80°C for up to 6 months, or at -20°C for up to 1 month, also protected from light.

  • Inhibits α-glucosidase
  • Isolated from natural plant sources
  • High purity of 99.79%
  • Appearance: White to yellow solid
  • Molecular weight: 610.52
  • Chemical formula: C27H30O16
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.