Learn More
Laboratory Equipment Co Panasonic MDF-U731M-PA Biomedical Freezer, -30°C Temperature Range, 24.4 cu.ft. Capacity
SDP
Catalog No. NC3653028
Encompass_Preferred
The Panasonic MDF-U731M-PA is a high-performance -30°C biomedical freezer designed for secure storage of sensitive biological materials. Its manual defrost system provides excellent temperature stability and uniformity, making it ideal for laboratories needing reliable preservation of blood components, enzymes, and vaccines. Key features include microprocessor control, comprehensive alarm functions, and a spacious 24.4 cu.ft. capacity, setting it apart from typical freezers in critical medical and research environments.
Catalog No. NC3653028 Supplier Laboratory Equipment Co Supplier No. MDFU731MPAELEMENT
Add to Cart
Add to Cart
Request a Quote
The Panasonic MDF-U731M-PA is a high-performance -30°C biomedical freezer designed for secure storage of sensitive biological materials. Its manual defrost system provides excellent temperature stability and uniformity, making it ideal for laboratories needing reliable preservation of blood components, enzymes, and vaccines. Key features include microprocessor control, comprehensive alarm functions, and a spacious 24.4 cu.ft. capacity, setting it apart from typical freezers in critical medical and research environments.