Learn More
Labrepco Inc PHCbi (formerly Panasonic) MDF Series 24.4 Cu. Ft. Manual Defrost Biomedical Freezer
SDP
Catalog No. NC1633383
Encompass_Preferred
The MDF-U731M-PA freezer is constructed with a high-performance clinical grade refrigeration system making it ideal for medical, biotechnology, and industrial laboratories storing blood components, enzymes, culture media, specimens, vaccines, and other temperature sensitive materials.
Catalog No. NC1633383 Supplier Labrepco Inc Supplier No. MDFU731MPA
Add to Cart
Add to Cart
Request a Quote
The MDF-U731M-PA freezer is constructed with a high-performance clinical grade refrigeration system making it ideal for medical, biotechnology, and industrial laboratories storing blood components, enzymes, culture media, specimens, vaccines, and other temperature sensitive materials.