Learn More
ATCC Panc 02.13; Pancreas Adenocarcinoma; Human (Homo sapiens)
Catalog No. 502383726
Encompass
Panc 02.03 is an epithelial cell line that was isolated in 1995 from the pancreas of a White, 70-year-old, female patient with adenocarcinoma. This cell line was deposited by EM Jaffee and can be used in cancer research.
Catalog No. 50-238-3726 Supplier ATCC Supplier No. CRL2554
Panc 02.03 is an epithelial cell line that was isolated in 1995 from the pancreas of a White, 70-year-old, female patient with adenocarcinoma. This cell line was deposited by EM Jaffee and can be used in cancer research.