Learn More
ATCC Panc 03.27; Pancreas Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2549
Encompass
Panc 03.27 is an epithelial cell line that was isolated in 1995 from the pancreas of a White, 65-year-old, female patient with adenocarcinoma. This cell line was deposited by EM Jaffee and can be used in cancer research.
Catalog No. 50-238-4390