Learn More
ATCC Panc 08.13; Pancreas Adenocarcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2551
Encompass
Panc 08.13 is an epithelial cell line that was isolated in 1995 from the pancreas of a White, 85-year-old, male patient with adenocarcinoma. This cell line was deposited by EM Jaffee and can be used in cancer research.
Catalog No. 50-238-4293