Learn More
ILEX LIFE SCIENCES LLC Immuchrom Pancreatic Elastase ELISA Kit for Human Stool Samples, 96 wells
SDP
Supplier:  ILEX LIFE SCIENCES LLC IC7400
Encompass_Preferred
The Immuchrom Pancreatic Elastase ELISA Kit is an immunoassay that enables easy, rapid and precise quantitative determination of pancreatic elastase in human stool samples. Calibration Range: 24.4 - 660 µg/ml. Detection Limit: 1.25 µg/ml. Principle: The pancreatic elastase test determines human pancreatic elastase according to the sandwich-principle. The elastase in sample, standard and controls binds to antibodies, which are coated to the microtiter plate. After a washing step a peroxidase labeled detection antibody is added. A second washing step is followed by the addition of the substrate which is converted to a colored product by the peroxidase. The reaction is terminated by adding an acidic stop solution. The optical densities are read at 450 nm (against the reference wavelength 620 nm) in a microtiter plate reader. The pancreatic-elastase concentration can be calculated from the standard curve. Synonyms: Pancreatic elastase-1, E1 enzyme, pancreatic endopeptidase E.
Catalog No. 50-212-0008