Learn More
ATCC PANC-1; Pancreatic Carcinoma; Human (Homo sapiens)
Catalog No. 502382594
Encompass
PANC-1 is a cell line exhibiting epithelial morphology that was isolated from the pancreatic duct of a 56-year-old, White, male with epithelioid carcinoma. This cell line can be used in cancer research.
Catalog No. 50-238-2594 Supplier ATCC Supplier No. CRL1469
PANC-1 is a cell line exhibiting epithelial morphology that was isolated from the pancreatic duct of a 56-year-old, White, male with epithelioid carcinoma. This cell line can be used in cancer research.