Learn More
ILEX LIFE SCIENCES LLC PANCOLL GRANU 1.119 20 X 500ML
SDP
Catalog No. 502733781
Encompass_Preferred
PAN-Biotech Pancoll Human for Granulocytes: Cell Separating Solution, Density: 1.119 g/ml (20 x 500 ml). MANUFACTURER: PAN-Biotech GmbH (Germany). STORAGE: +2°C-RT. DENSITY: 1.119 g/ml. EDTA: Without EDTA. HEPES: Without HEPES. STERILE: Yes. DESCRIPTION: Density gradient centrifugation media to easily isolate granulocytes from red blood cells! In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose so-called separating solutions (also known as centrifugation media or density gradient media) are used. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell separation applications.
Catalog No. 50-273-3781 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0460150X20
PAN-Biotech Pancoll Human for Granulocytes: Cell Separating Solution, Density: 1.119 g/ml (20 x 500 ml). MANUFACTURER: PAN-Biotech GmbH (Germany). STORAGE: +2°C-RT. DENSITY: 1.119 g/ml. EDTA: Without EDTA. HEPES: Without HEPES. STERILE: Yes. DESCRIPTION: Density gradient centrifugation media to easily isolate granulocytes from red blood cells! In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose so-called separating solutions (also known as centrifugation media or density gradient media) are used. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell separation applications.