Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech Pancoll human, Cell Separating Solution (Centrifugation Media), Density: 1.100 g/ml (500 ml)
SDP
Catalog No. 502388471
Encompass_Preferred
PAN-Biotech Pancoll Human: Cell Separating Solution (centrifugation media), Density: 1.100 g/ml, quantity: 500 ml. DESCRIPTION: In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. Separating solutions are used for this purpose. Pancoll separating solutions from PAN-Biotech contain a polysaccharide with a molecular weight of 400,000 daltons. This hydrophilic polymer allows production of aqueous solutions for cell separation with a density of up to 1.119 g/ml. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell separation applications. DENSITY: 1.100 g/ml. EDTA: without EDTA. HEPES: without HEPES. STERILE: yes. STORAGE: +2°C - RT. COUNTRY OF ORIGIN: Germany.
Catalog No. 50-238-8471 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0469600
PAN-Biotech Pancoll Human: Cell Separating Solution (centrifugation media), Density: 1.100 g/ml, quantity: 500 ml. DESCRIPTION: In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. Separating solutions are used for this purpose. Pancoll separating solutions from PAN-Biotech contain a polysaccharide with a molecular weight of 400,000 daltons. This hydrophilic polymer allows production of aqueous solutions for cell separation with a density of up to 1.119 g/ml. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell separation applications. DENSITY: 1.100 g/ml. EDTA: without EDTA. HEPES: without HEPES. STERILE: yes. STORAGE: +2°C - RT. COUNTRY OF ORIGIN: Germany.