Learn More
ILEX LIFE SCIENCES LLC PANCOLL PRO BLOOD CELL 500 ML
SDP
Catalog No. 502733779
Encompass_Preferred
PAN-Biotech Pancoll Pro, Blood Cell Separation Medium, Low Endotoxin (<0.1 EU/ml), Density 1.077 g/ml, 500 ml. DENSITY: 1.077 g/ml. MANUFACTURER: PAN-Biotech GmbH (Germany). STORAGE: +2°C-RT. EDTA: Without EDTA. HEPES: Without HEPES. STERILE: Yes. COMPOSITION: Polysucrose: ~ 57,000.00 mg/L; Diatrizoic Acid: ~ 86,900.00 mg/L. DESCRIPTION: In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose so-called separation solutions (also known as centrifugation media or density gradient media) are used. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell isolation applications.
Catalog No. 50-273-3779 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0460505
Add to Cart
Add to Cart
PAN-Biotech Pancoll Pro, Blood Cell Separation Medium, Low Endotoxin (<0.1 EU/ml), Density 1.077 g/ml, 500 ml. DENSITY: 1.077 g/ml. MANUFACTURER: PAN-Biotech GmbH (Germany). STORAGE: +2°C-RT. EDTA: Without EDTA. HEPES: Without HEPES. STERILE: Yes. COMPOSITION: Polysucrose: ~ 57,000.00 mg/L; Diatrizoic Acid: ~ 86,900.00 mg/L. DESCRIPTION: In many cases the isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose so-called separation solutions (also known as centrifugation media or density gradient media) are used. PAN-Biotech offers a variety of ready-to-use products with a density of 1.063 g/ml up to 1.119 g/ml for a very wide range of cell isolation applications.