Learn More
CPC Scientific H-Glu-Glu-Glu-Glu-Glu-Met-Ala-Val-Val-Pro-Gln-Gly-Leu-Phe-Arg-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PANS001A
Encompass
Pancreastatin strongly inhibits glucose-induced and gastric inhibitory peptide-induced insulin secretion from rat pancreas. It shows a high sequence homology to the central part of bovine, human, and rat chromogranin A.ONE-LETTER SEQUENCE: EEEEEMAVVPQGLFRGMOLECULAR FORMULA: C78H123N21O27S1MOLECULAR WEIGHT:1819.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [133605-57-9], RESEARCH AREA: DiabetesREFERENCES: O?Connor et al., Endocrinol Metal 90(9):5414-25 (2005)
Catalog No. 50-211-0085