Learn More
NEOGEN Pancreatic Digest of Gelatin, 5 kg
SDP
Catalog No. 502015553
Encompass_Preferred
Pancreatic Digest of Gelatin is an enzymatic digest of gelatin for use in preparing microbiological culture media in a laboratory setting. Pancreatic Digest of Gelatin is not intended for use in the diagnosis of disease or other conditions in humans.Pancreatic Digest of Gelatin provides a complex nitrogen source and is used as a media ingredient for fermentation studies. Pancreatic Digest of Gelatin alone is used to support the growth of non-fastidious microorganisms. Pancreatic Digest of Gelatin is deficient in carbohydrates and distinguished by low cystine and tryptophan content.For more information visit https://www.neogen.com/solutions/Microbiology/pancreatic-digest-gelatin
Catalog No. 50-201-5553 Supplier NEOGEN Supplier No. 700003369
Pancreatic Digest of Gelatin is an enzymatic digest of gelatin for use in preparing microbiological culture media in a laboratory setting. Pancreatic Digest of Gelatin is not intended for use in the diagnosis of disease or other conditions in humans.Pancreatic Digest of Gelatin provides a complex nitrogen source and is used as a media ingredient for fermentation studies. Pancreatic Digest of Gelatin alone is used to support the growth of non-fastidious microorganisms. Pancreatic Digest of Gelatin is deficient in carbohydrates and distinguished by low cystine and tryptophan content.For more information visit https://www.neogen.com/solutions/Microbiology/pancreatic-digest-gelatin