Learn More
Medchemexpress LLC Pancreatic lipase/carboxylesterase 1-IN-1 | 194235-21-7 | 99.9% | 468.67 | C30H44O4 | 5 MG
SDP
Catalog No. 5000221306
Encompass_Preferred
Pancreatic lipase/Carboxylesterase 1-IN-1 is a research-grade small-molecule inhibitor active against pancreatic lipase and human carboxylesterase 1A. Supplied as a solid for biochemical and cellular assays, it has reported potency values against both enzymes and is intended for laboratory research use only.
  • Dual inhibitor of pancreatic lipase and human carboxylesterase 1A
  • Reported IC50: 2.13 μM (pancreatic lipase), 0.055 μM (hCES1A)
  • Molecular formula C30H44O4 and molecular weight 468.67
  • High purity (99.9%) suitable for biochemical assays
  • Available in small package sizes such as 5 mg and 10 mg for research use
  • White to off-white solid; follow SDS for storage and handling
Catalog No. 50-002-21306 Supplier Medchemexpress LLC Supplier No. HY1448095MG
Pancreatic lipase/Carboxylesterase 1-IN-1 is a research-grade small-molecule inhibitor active against pancreatic lipase and human carboxylesterase 1A. Supplied as a solid for biochemical and cellular assays, it has reported potency values against both enzymes and is intended for laboratory research use only.
  • Dual inhibitor of pancreatic lipase and human carboxylesterase 1A
  • Reported IC50: 2.13 μM (pancreatic lipase), 0.055 μM (hCES1A)
  • Molecular formula C30H44O4 and molecular weight 468.67
  • High purity (99.9%) suitable for biochemical assays
  • Available in small package sizes such as 5 mg and 10 mg for research use
  • White to off-white solid; follow SDS for storage and handling