Learn More
Medchemexpress LLC Pancreatic lipase/carboxylesterase 1-in-1 | 194235-21-7 | 99.9% | C30H44O4 | 10MG
SDP
Catalog No. 5000258409
Encompass_Preferred
Pancreatic lipase/Carboxylesterase 1-IN-1 is a small-molecule dual inhibitor of pancreatic lipase and human carboxylesterase 1 (hCES1) supplied for research use. It is a white to off-white solid with chemical formula C30H44O4 and molecular weight 468.67 g·mol⁻1, provided in small milligram pack sizes for laboratory assays.
  • Dual inhibitor of pancreatic lipase and human carboxylesterase 1 (hCES1).
  • Chemical formula C30H44O4 and molecular weight 468.67 g·mol⁻1.
  • High listed purity (99.9%).
  • Supplied as a white to off-white solid suitable for laboratory use.
  • Intended for research use only; not for human use.
  • Available in small milligram pack sizes for assay development.
Catalog No. 50-002-58409 Supplier Medchemexpress LLC Supplier No. HY14480910MG
Pancreatic lipase/Carboxylesterase 1-IN-1 is a small-molecule dual inhibitor of pancreatic lipase and human carboxylesterase 1 (hCES1) supplied for research use. It is a white to off-white solid with chemical formula C30H44O4 and molecular weight 468.67 g·mol⁻1, provided in small milligram pack sizes for laboratory assays.
  • Dual inhibitor of pancreatic lipase and human carboxylesterase 1 (hCES1).
  • Chemical formula C30H44O4 and molecular weight 468.67 g·mol⁻1.
  • High listed purity (99.9%).
  • Supplied as a white to off-white solid suitable for laboratory use.
  • Intended for research use only; not for human use.
  • Available in small milligram pack sizes for assay development.