Learn More
Eurofins Discoverx Pancreatic Polypeptide, Human, 0.5 mg. (92-1062)
Supplier:  Eurofins Discoverx 921062
Encompass
Pancreatic polypeptide is a 95 amino acid polypeptide secreted by the pancreas that is cleaved into a 36 amino acid active hormone that regulates pancreatic secretion, and an icosapeptide of unknown function. It is an endogenous agonist of the NPY Y4 receptor PPYR1 (Ki=0.056 nM), and may play an important part in gastrointestinal function. Provided as 500 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9603