Learn More
Bachem Pancreatic Polypeptide (rat)
SDP
Supplier:  Bachem 4031085.0500
Encompass_Preferred
Pancreatic Polypeptide (rat), H-Ala-Pro-Leu-Glu-Pro-Met-Tyr-Pro-Gly-Asp-Tyr-Ala-Thr-His-Glu-Gln-Arg-Ala-Gln-Tyr-Glu-Thr-Gln-Leu-Arg-Arg-Tyr-Ile-Asn-Thr-Leu-Thr-Arg-Pro-Arg-Tyr-NH2, 90419-12-8, C195H298N58O57S, Mr 4398.93, 0.5mg. The sequence also corresponds to the mouse hormone.
Catalog No. 50-260-130