Learn More
Bachem Pancreatic Polypeptide (human) Trifluoroacetate
SDP
Supplier:  Bachem 4015101.0500
Encompass_Preferred
Pancreatic Polypeptide (human) Trifluoroacetate, H-Ala-Pro-Leu-Glu-Pro-Val-Tyr-Pro-Gly-Asp-Asn-Ala-Thr-Pro-Glu-Gln-Met-Ala-Gln-Tyr-Ala-Ala-Asp-Leu-Arg-Arg-Tyr-Ile-Asn-Met-Leu-Thr-Arg-Pro-Arg-Tyr-NH2, 75976-10-2, C185H287N53O54S2, Mr 4181.77, 0.5mg. As antagonist of CCK, PP inhibits pancreatic sedretion. PP also acts on the gastrointestinal motility, food intake, and metabolism.
Catalog No. NC1249604