Learn More
Diagnostic Biosystems PANCREATIC POLYPEPTIDE Pancreatic Polypeptide
Supplier:  Diagnostic Biosystems RP030
Encompass_Preferred
This antibody reacts with pancreatic polypeptide. It stains the periphery of islets, exocrine pancreatic parenchyma and the epithelium of small and medium sized ducts and acinar cells.
Catalog No. 50-238-5456