Learn More
Echelon Biosciences Research Labs Pancreatic Polypeptide, avian - 0.5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 478400.5MG
Encompass_Preferred
Pancreatic Polypeptide, avian - 0.5mg , Gly-Pro-Ser-Gln-Pro-Thr-Tyr-Pro-Gly-Asp-Asp-Ala-Pro-Val-Glu-Asp-Leu-Ile-Arg-Phe-Tyr-Asp-Asn-Leu-Gln-Gln-Tyr-Leu-Asn-Val-Val-Thr-Arg-His-Arg-Tyr-NH2 This item is non returnable
Catalog No. 50-190-8538