Learn More
Medchemexpress LLC HY-B2118 500mg Medchemexpress, Pancreatin CAS:8049-47-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2118500MG
Encompass_Preferred
Medchemexpress, HY-B2118 500mg Pancreatin CAS:8049-47-6 Pancreatin is the porcine pancreas extract (PPE) which contains the main pancreatic digestive enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2723