Learn More
Creative Biomart Pancrelipase protease
Supplier:  Creative Biomart THP0041
Encompass_Preferred
Pancrelipase, in general, is composed of a mixture of pancreatic enzymes which include amylases, lipases, and proteases. These enzymes are extracted from porcine pancreatic glands. The product is a mix of enzymes, formed by trypsin and chymotrypsin, that proteolytically cleave peptide bonds and are involved in food digestion.
Catalog No. 50-208-4092