Learn More
AdipoGen Pancuronium bromide (100 mg)
SDP
Supplier:  AdipoGen CDXP0238M100
Encompass_Preferred
Chemical. CAS: 15500-66-0. Formula: C35H60Br2N2O4. MW: 732.67. Pancuronium bromide is an aminosteroid antagonist of muscle-type nicotinic acetylcholine receptors (nAChRs) acting as a typical non-depolarizing curare-mimetic muscle relaxant/neuromuscular blocking agent. Pancuronium bromide enhances anesthesia induced by isoflurane.
Catalog No. 50-205-4595