Learn More
Selleck Chemical LLC Pancuronium dibromide S2497-1g
SDP
Catalog No. 503111634
Encompass_Preferred
Pancuronium dibromide is a competitive nicotinic acetylcholine receptor antagonist used as a muscle relaxant
Catalog No. 50-311-1634 Supplier Selleck Chemical LLC Supplier No. S24971G
Pancuronium dibromide is a competitive nicotinic acetylcholine receptor antagonist used as a muscle relaxant