Learn More
Selleck Chemical LLC Pancuronium dibromide S2497-200mg
SDP
Catalog No. 503111751
Encompass_Preferred
Pancuronium dibromide is a competitive nicotinic acetylcholine receptor antagonist used as a muscle relaxant
Catalog No. 50-311-1751 Supplier Selleck Chemical LLC Supplier No. S2497200MG
Pancuronium dibromide is a competitive nicotinic acetylcholine receptor antagonist used as a muscle relaxant