Learn More
Medchemexpress LLC HY-B0429 10mg Medchemexpress, Pancuronium (dibromide) CAS:15500-66-0 Purity:>98%
SDP
Catalog No. 502082532
Encompass_Preferred
Medchemexpress, HY-B0429 10mg Pancuronium (dibromide) CAS:15500-66-0 Pancuronium dibromide, a bis-quaternary steroid, is a neuromuscular relaxant. Pancuronium dibromide inhibits neuromuscular transmission by competing with acetylcholine for binding sites on nACh receptors. Pancuronium dibromide also inhibits cardiac muscarinic receptors and has a sympathomimetic action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2532 Supplier Medchemexpress LLC Supplier No. HYB042910MG
Medchemexpress, HY-B0429 10mg Pancuronium (dibromide) CAS:15500-66-0 Pancuronium dibromide, a bis-quaternary steroid, is a neuromuscular relaxant. Pancuronium dibromide inhibits neuromuscular transmission by competing with acetylcholine for binding sites on nACh receptors. Pancuronium dibromide also inhibits cardiac muscarinic receptors and has a sympathomimetic action. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.