Learn More
Apexbio Technology LLC Pancuronium dibromide 15500-66-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063383
Encompass_Preferred
Pancuronium dibromide (CAS 15500-66-0) is a small molecule that acts as a competitive antagonist at nicotinic acetylcholine receptors (AChRs) blocking neuromuscular transmission by displacing acetylcholine from its binding sites on the motor end-plate This action results in skeletal muscle relaxation characterized by dose-dependent neuromuscular blockade With an IC50 value of approximately 5 5 nM pancuronium dibromide serves as a potent pharmacological tool for studying cholinergic receptor function neuromuscular signaling pathways and muscle physiology in biomedical research
Catalog No. 50-000-63383 Supplier Apexbio Technology LLC Supplier No. B16125.1
Pancuronium dibromide (CAS 15500-66-0) is a small molecule that acts as a competitive antagonist at nicotinic acetylcholine receptors (AChRs) blocking neuromuscular transmission by displacing acetylcholine from its binding sites on the motor end-plate This action results in skeletal muscle relaxation characterized by dose-dependent neuromuscular blockade With an IC50 value of approximately 5 5 nM pancuronium dibromide serves as a potent pharmacological tool for studying cholinergic receptor function neuromuscular signaling pathways and muscle physiology in biomedical research