Learn More
TARGETMOL CHEMICALS INC Pancuronium dibromide 50MG
SDP
Catalog No. 502880566
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Pancuronium dibromide (Pavulon), a competitive AChR antagonist (IC50 = 5.5 nM), acts as a skeletal muscle relaxant. Pancuronium dibromide(Pancuronium bromide) blocking neuromuscular transmission is achieved by competing with acetylcholine for receptor sites on the motor end-plate. Purity 99.27%
Catalog No. 50-288-0566 Supplier TARGETMOL CHEMICALS INC Supplier No. T016650MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Pancuronium dibromide (Pavulon), a competitive AChR antagonist (IC50 = 5.5 nM), acts as a skeletal muscle relaxant. Pancuronium dibromide(Pancuronium bromide) blocking neuromuscular transmission is achieved by competing with acetylcholine for receptor sites on the motor end-plate. Purity 99.27%