Learn More
Cable Ties And More PANDUIT POLY CABLE 11.5"1000CS
SDP
Catalog No. NC1917325
Encompass
Panduit® Polypropylene Cable Ties 11.5" long, 30 lbs strength. 1000 pcs per pack. Weather Resistant. Operating Temperature: -60ºC To +150ºC
Catalog No. NC1917325 Supplier Cable Ties And More Supplier No. PLT3SM100
Panduit® Polypropylene Cable Ties 11.5" long, 30 lbs strength. 1000 pcs per pack. Weather Resistant. Operating Temperature: -60ºC To +150ºC