Learn More
Cable Ties And More PANDUIT POLY CABLE 7", 1000CS
SDP
Catalog No. NC1917319
Encompass
Panduit® Polypropylene Cable Ties 7" long, 30bs strength. 1000 pcs per pack. black.Weather Resistant.Operating Temperature: -60ºC To +150ºCOperating Temperature: -60ºC To +150ºC
Catalog No. NC1917319 Supplier Cable Ties And More Supplier No. PLT2SM100
Panduit® Polypropylene Cable Ties 7" long, 30bs strength. 1000 pcs per pack. black.Weather Resistant.Operating Temperature: -60ºC To +150ºCOperating Temperature: -60ºC To +150ºC