Learn More
Medchemexpress LLC Panduratin A | 89837-52-5 | 406.51 | 1 MG
SDP
Catalog No. 5000415726
Encompass_Preferred
Panduratin A is an orally active natural compound known for its diverse pharmacological activities. It functions as a potent anti-inflammatory and antioxidant agent by specifically inhibiting the NF-κB signaling pathway in models of intestinal and vascular inflammation. It also offers protection against Colistin-induced nephrotoxicity and activates autophagy through an AMPK-dependent pathway, demonstrating anti-tuberculosis and antiviral effects.
  • Anti-inflammatory and antioxidant effects
  • Nephroprotective properties
  • Autophagy activation
  • Anti-tuberculosis activity
  • Antiviral effects by inhibiting SARS-CoV-2 methyltransferase
  • High purity of 99.9%
Catalog No. 50-004-15726 Supplier Medchemexpress LLC Supplier No. HYN117221MG
Panduratin A is an orally active natural compound known for its diverse pharmacological activities. It functions as a potent anti-inflammatory and antioxidant agent by specifically inhibiting the NF-κB signaling pathway in models of intestinal and vascular inflammation. It also offers protection against Colistin-induced nephrotoxicity and activates autophagy through an AMPK-dependent pathway, demonstrating anti-tuberculosis and antiviral effects.
  • Anti-inflammatory and antioxidant effects
  • Nephroprotective properties
  • Autophagy activation
  • Anti-tuberculosis activity
  • Antiviral effects by inhibiting SARS-CoV-2 methyltransferase
  • High purity of 99.9%