Learn More
MSC Baldwin Filters Panel Air Element, Heavy Duty, 20 in., 1 ea
Catalog No. 5000509848
Encompass
This panel heavy-duty air element is designed to provide efficient filtration for engine air intake systems, ensuring optimal performance and protection against contaminants. It is specifically engineered to fit John Deere equipment and Elgin sweepers.
  • Features an attached gasket for a secure seal
  • Constructed as a reliable panel-style air element
  • Designed for high-performance heavy-duty applications
Catalog No. 50-005-09848 Supplier MSC Supplier No. 55873905
This panel heavy-duty air element is designed to provide efficient filtration for engine air intake systems, ensuring optimal performance and protection against contaminants. It is specifically engineered to fit John Deere equipment and Elgin sweepers.
  • Features an attached gasket for a secure seal
  • Constructed as a reliable panel-style air element
  • Designed for high-performance heavy-duty applications