Learn More
MSC Baldwin Filters, PANEL HD AIR ELEMENT
Catalog No. 5000508306
Encompass
These panel air elements are designed to protect vehicles and heavy equipment by efficiently removing airborne contaminants from the engine intake air. Engineered for performance, these filters provide robust filtration to maintain optimal engine health and operational reliability.
  • Designed for effective removal of airborne contaminants
  • Engine air filter with panel design
  • Constructed for engine and automotive applications
  • Dimensions: 508 mm x 381 mm x 55.6 mm (20 in. x 15 in. x 2 3/16 in.)
Catalog No. 50-005-08306 Supplier MSC Supplier No. 55869366
These panel air elements are designed to protect vehicles and heavy equipment by efficiently removing airborne contaminants from the engine intake air. Engineered for performance, these filters provide robust filtration to maintain optimal engine health and operational reliability.
  • Designed for effective removal of airborne contaminants
  • Engine air filter with panel design
  • Constructed for engine and automotive applications
  • Dimensions: 508 mm x 381 mm x 55.6 mm (20 in. x 15 in. x 2 3/16 in.)