Learn More
Grainger Panel Mount Block Anodized Aluminum
Catalog No. 502766465
Encompass_Preferred
The 80/20 Panel Mount Block for Standard T-Slotted Framing is a versatile mounting solution for attaching panels or other components to 80/20 aluminum framing. This black anodized aluminum block has two T-nuts that slide into the T-slots on 80/20 framing, allowing the block to be securely mounted. It features 4 counterbored holes that accept 1/4"-20 or M6 bolts, providing multiple mounting options. With its modular design, this panel mount block can be used to attach panels, plates, brackets, or other components to 80/20 framing quickly and easily.
Catalog No. 50-276-6465 Supplier Grainger Supplier No. 16U341
The 80/20 Panel Mount Block for Standard T-Slotted Framing is a versatile mounting solution for attaching panels or other components to 80/20 aluminum framing. This black anodized aluminum block has two T-nuts that slide into the T-slots on 80/20 framing, allowing the block to be securely mounted. It features 4 counterbored holes that accept 1/4"-20 or M6 bolts, providing multiple mounting options. With its modular design, this panel mount block can be used to attach panels, plates, brackets, or other components to 80/20 framing quickly and easily.