Learn More
Indigo Biosciences Human LXR Reporter Assays Panel All-inclusive Cell-Based Luciferase Reporter Assay Kit, Including LXRa & LXRb, each in 1x48 96-well format
Supplier:  Indigo Biosciences IB0032148P
Encompass_Preferred
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Human Liver X Receptor Panel (LXR alpha, LXR beta) assay kit is an all-inclusive firefly luciferase reporter assay system that includes in addition to reporter cells for each LXR, optimized growth media, media for diluting test compounds, a control agonist for each LXR, luciferase detection reagent, a cell culture-ready assay plate in strip-well format so that (if preferred) LXR alpha and LXR beta assays may be performed at different times, detailed protocol, and a protocol quick guide. INDIGO’s unique Cryomite™ process provides exceptional cell viability post-thaw and INDIGO’s strict QA process means consistent results lot to lot (product quality data available upon request). This kit allows researchers to achieve superior sensitivity, clear, reproducible results, and eliminate weeks of cell culture work.
Catalog No. NC3588542