Learn More
Grainger Panel Pilot Cut Profile Router Bit 1/2
Catalog No. 502766900
Encompass_Preferred
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative profiles, grooves, and edges into wood, plastic, and non-ferrous metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-22,000 RPM. The router comes with a 1/4" and 1/2" collet, dust extraction adapter, edge guide, wrenches, and plastic carrying case. This versatile router allows for plunge cuts and use with templates. It weighs 6 lbs and has soft grip handles for comfort and control. The Panel Pilot router is ideal for sign makers, woodworkers, and hobbyists looking to add decorative details.
Catalog No. 50-276-6900 Supplier Grainger Supplier No. 16Y622
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative profiles, grooves, and edges into wood, plastic, and non-ferrous metals. It has a 1-1/4 HP motor with a variable speed range of 10,000-22,000 RPM. The router comes with a 1/4" and 1/2" collet, dust extraction adapter, edge guide, wrenches, and plastic carrying case. This versatile router allows for plunge cuts and use with templates. It weighs 6 lbs and has soft grip handles for comfort and control. The Panel Pilot router is ideal for sign makers, woodworkers, and hobbyists looking to add decorative details.