Learn More
Grainger Panel Pilot Cut Profile Router Bit 1/4
Catalog No. 502765218
Encompass_Preferred
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative edge profiles, circles, and other shapes in wood, plastic, and soft metals. It has a 1-1/4 HP motor with variable speed from 10,000 to 23,000 RPM. The router accepts Porter-Cable style template guides and 1/4" and 1/2" collets. It comes with a plastic sub-base, chip deflector, wrenches, collets, and a user manual. This versatile router is ideal for sign making, woodworking, plastic fabrication, light metal work, and other routing applications requiring precise edge profiles.
Catalog No. 50-276-5218 Supplier Grainger Supplier No. 16Y532
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative edge profiles, circles, and other shapes in wood, plastic, and soft metals. It has a 1-1/4 HP motor with variable speed from 10,000 to 23,000 RPM. The router accepts Porter-Cable style template guides and 1/4" and 1/2" collets. It comes with a plastic sub-base, chip deflector, wrenches, collets, and a user manual. This versatile router is ideal for sign making, woodworking, plastic fabrication, light metal work, and other routing applications requiring precise edge profiles.