Learn More
Grainger Panel Pilot Cut Profile Router Bit 1/2
Catalog No. 502765238
Encompass_Preferred
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative profiles, grooves, and edges into wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed dial that provides 10,000 to 23,000 RPM. The router comes with a 1/4-inch and 1/2-inch collet, dust extraction adapter, edge guide, wrenches, and plastic case. This versatile, compact router is ideal for sign making, woodworking, hobby, and light-duty routing applications.
Catalog No. 50-276-5238 Supplier Grainger Supplier No. 16Y619
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative profiles, grooves, and edges into wood, plastic, and soft metals. It has a 1-1/4 HP motor with a variable speed dial that provides 10,000 to 23,000 RPM. The router comes with a 1/4-inch and 1/2-inch collet, dust extraction adapter, edge guide, wrenches, and plastic case. This versatile, compact router is ideal for sign making, woodworking, hobby, and light-duty routing applications.