Learn More
Grainger Panel Pilot Cut Profile Router Bit 1/2
Catalog No. 502766862
Encompass_Preferred
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative edge profiles, circles, and other shapes in wood, plastic, and soft metals. It has a 1-1/4 HP motor with variable speed from 10,000 to 23,000 RPM. The router accepts Porter-Cable style template guides and 1/4" and 1/2" collets. It comes with a 1/4" and 1/2" collet, edge guide, wrenches, and dust extraction adapter. This versatile router allows for handheld use or mounting in a router table. It's compact and lightweight at only 4.2 lbs.
Catalog No. 50-276-6862 Supplier Grainger Supplier No. 16Y539
The WESTWARD Panel Pilot Cut Profile Router is a handheld power tool designed for cutting decorative edge profiles, circles, and other shapes in wood, plastic, and soft metals. It has a 1-1/4 HP motor with variable speed from 10,000 to 23,000 RPM. The router accepts Porter-Cable style template guides and 1/4" and 1/2" collets. It comes with a 1/4" and 1/2" collet, edge guide, wrenches, and dust extraction adapter. This versatile router allows for handheld use or mounting in a router table. It's compact and lightweight at only 4.2 lbs.