Learn More
Grainger 80/20 Inc. Panel Retainer, 15 Series, Aluminum, Anodized, 6/Pk.
Catalog No. 502766478
Encompass_Preferred
This single arm wide panel retainer is designed for mounting panels that are recessed within a T-slot or on a profile frame, providing a stable, vibration-reducing connection for 15 Series and Ready Tube systems. It is suitable for applications where a single panel is mounted to a profile or when larger profile series are used to secure two panels.
  • Features a robust 6063-T6 aluminum construction with an anodized finish.
  • Requires drill-through machining service on the panel for installation.
  • Designed for 15 Series and Ready Tube profiles.
  • Provides 4 mounting holes with 5/16-18 tapped threads.
  • ESD safe design suitable for sensitive environments.
Catalog No. 50-276-6478 Supplier Grainger Supplier No. 16U339
This single arm wide panel retainer is designed for mounting panels that are recessed within a T-slot or on a profile frame, providing a stable, vibration-reducing connection for 15 Series and Ready Tube systems. It is suitable for applications where a single panel is mounted to a profile or when larger profile series are used to secure two panels.
  • Features a robust 6063-T6 aluminum construction with an anodized finish.
  • Requires drill-through machining service on the panel for installation.
  • Designed for 15 Series and Ready Tube profiles.
  • Provides 4 mounting holes with 5/16-18 tapped threads.
  • ESD safe design suitable for sensitive environments.