Learn More
Grainger Panel Retainer Anodized Aluminum
Catalog No. 502766834
Encompass_Preferred
The 80/20 Panel Retainer for Standard Panels is a metal panel retainer designed for use with 80/20 aluminum framing systems. It is made from 6061-T6 aluminum and has a black anodized finish. The panel retainer measures 0.625" wide x 0.375" thick x 1.5" long and features a spring-loaded design to hold panels securely in place. It can accommodate panel thicknesses from 0.080" to 0.250". The panel retainer allows for easy installation and removal of panels on 80/20 framing without requiring tools or fasteners. It provides a clean, simple method to mount and change panels on 80/20 aluminum extrusion frames.
Catalog No. 50-276-6834 Supplier Grainger Supplier No. 16U344
The 80/20 Panel Retainer for Standard Panels is a metal panel retainer designed for use with 80/20 aluminum framing systems. It is made from 6061-T6 aluminum and has a black anodized finish. The panel retainer measures 0.625" wide x 0.375" thick x 1.5" long and features a spring-loaded design to hold panels securely in place. It can accommodate panel thicknesses from 0.080" to 0.250". The panel retainer allows for easy installation and removal of panels on 80/20 framing without requiring tools or fasteners. It provides a clean, simple method to mount and change panels on 80/20 aluminum extrusion frames.