Learn More
Grainger 80/20 Panel Retainer, Aluminum, 15 Series, Silver, 1 ea
Catalog No. 502763738
Encompass_Preferred
This panel retainer is designed for use with 15 series T-slot framing systems. Constructed from 6063-T6 aluminum with an anodized finish, this component provides a secure mounting solution for standard panels.
  • Manufactured from 6063-T6 aluminum
  • Anodized finish for durability
  • Compatible with 15 series framing systems
  • Features two mounting holes with 21/64 in. diameter
  • Dimensions measure 1 1/2 in. long by 1 1/2 in. wide by 27/32 in. high
  • ESD safe construction
  • Designed for 0.4 in. slot width
  • Requires 5/16 in. fasteners (not included)
Catalog No. 50-276-3738 Supplier Grainger Supplier No. 16U365
This panel retainer is designed for use with 15 series T-slot framing systems. Constructed from 6063-T6 aluminum with an anodized finish, this component provides a secure mounting solution for standard panels.
  • Manufactured from 6063-T6 aluminum
  • Anodized finish for durability
  • Compatible with 15 series framing systems
  • Features two mounting holes with 21/64 in. diameter
  • Dimensions measure 1 1/2 in. long by 1 1/2 in. wide by 27/32 in. high
  • ESD safe construction
  • Designed for 0.4 in. slot width
  • Requires 5/16 in. fasteners (not included)