Learn More
Hach Company Panel- SC4500-CL17sc-pH
Catalog No. 5000207724 Shop All Hach Company Products
Encompass
Dual Parameter Monitoring Panel with SC4500-CL17sc and digital pH SensorThe Dual Parameter Chlorine panels offers CL17sc chlorine analyzer Digital pH sensor and SC4500 controller in the convenience of one panel These panel configurations are pre-engineered ready to install and simple solutions that are designed to work together
Catalog No. 50-002-07724 Supplier Hach Company Supplier No. DPMPCLPHD
Add to Cart
Add to Cart
Request a Quote
Dual Parameter Monitoring Panel with SC4500-CL17sc and digital pH SensorThe Dual Parameter Chlorine panels offers CL17sc chlorine analyzer Digital pH sensor and SC4500 controller in the convenience of one panel These panel configurations are pre-engineered ready to install and simple solutions that are designed to work together