Learn More
Hach Company Panel- SC4500-TU5300sc-pH
Supplier:  Hach Company DPMPTUPHD
Encompass
Dual Parameter Monitoring Panel with SC4500-TU5300sc and digital pH SensorThe Dual Parameter Turbidity panels offer TU5 Series TU5300sc Low Range Laser Turbidimeter Digital pH Sensor and SC4500 controller in the convenience of one panel These panel configurations are pre-engineered ready to install simple solutions that are designed to work together
Catalog No. 50-002-07552
Add to Cart
Request a Quote