Learn More
Grainger Panel Truck 3200 lb Cap 54inL 30inW
Catalog No. 502751726
Encompass_Preferred
The Raymond Products Multiheight Vertical Panel Saw is a vertical panel saw designed for cutting large panels and sheets. It features a multiheight cutting capacity of up to 4 inches and can cut materials up to 5 feet wide. The saw has a 3 HP motor with electronic variable speed control from 1800-4000 RPM. It includes a digital protractor display for precise angle cutting from -45 to +45 degrees. The saw table is precision ground cast iron with a durable powder coat finish. It comes with an adjustable rip fence with micro adjustment. This industrial grade vertical panel saw is made in the USA.
Catalog No. 50-275-1726 Supplier Grainger Supplier No. 14G819
The Raymond Products Multiheight Vertical Panel Saw is a vertical panel saw designed for cutting large panels and sheets. It features a multiheight cutting capacity of up to 4 inches and can cut materials up to 5 feet wide. The saw has a 3 HP motor with electronic variable speed control from 1800-4000 RPM. It includes a digital protractor display for precise angle cutting from -45 to +45 degrees. The saw table is precision ground cast iron with a durable powder coat finish. It comes with an adjustable rip fence with micro adjustment. This industrial grade vertical panel saw is made in the USA.