Learn More
Grainger Panel Truck 3200 lb Cap 54inL 30inW
Catalog No. 502751772
Encompass_Preferred
The Raymond Products Multiheight Vertical Panel Saw is a vertical panel saw designed for cutting large panels. It features a multiheight cutting capacity of up to 4 feet for cutting plywood, melamine, plastic laminates and more. The saw has a powerful 5HP motor with electronic variable speed from 0 to 4000 RPM. It includes a digital protractor for precise angle cuts from -45 to +45 degrees. The saw table is made from precision ground cast iron for durability. Safety features include an electronic brake and blade guard. This industrial-grade vertical panel saw is ideal for cabinet shops, millwork operations, and other woodworking applications requiring accurate cuts on large panels.
Catalog No. 50-275-1772 Supplier Grainger Supplier No. 14G814
The Raymond Products Multiheight Vertical Panel Saw is a vertical panel saw designed for cutting large panels. It features a multiheight cutting capacity of up to 4 feet for cutting plywood, melamine, plastic laminates and more. The saw has a powerful 5HP motor with electronic variable speed from 0 to 4000 RPM. It includes a digital protractor for precise angle cuts from -45 to +45 degrees. The saw table is made from precision ground cast iron for durability. Safety features include an electronic brake and blade guard. This industrial-grade vertical panel saw is ideal for cabinet shops, millwork operations, and other woodworking applications requiring accurate cuts on large panels.