Learn More
Grainger Panel Truck 3200 lb Cap 54inL 30inW
Catalog No. 502753643
Encompass_Preferred
The Grainger 14G824 is a compact, portable air compressor that delivers 2.0 SCFM at 90 PSI with a maximum pressure of 135 PSI. It has a 1/4 horsepower oil-free pump and is powered by a standard 120V household outlet. This air compressor includes a 6 foot power cord and measures 9-1/2 inches high by 11-1/2 inches wide by 10-1/2 inches deep. It weighs 16 pounds and has a 1 gallon steel tank with manual drain. The 14G824 can be used for inflation, stapling, brad nailing, air brushing and other light duty applications. This is an oil-free piston type compressor from Grainger.
Catalog No. 50-275-3643 Supplier Grainger Supplier No. 14G824
The Grainger 14G824 is a compact, portable air compressor that delivers 2.0 SCFM at 90 PSI with a maximum pressure of 135 PSI. It has a 1/4 horsepower oil-free pump and is powered by a standard 120V household outlet. This air compressor includes a 6 foot power cord and measures 9-1/2 inches high by 11-1/2 inches wide by 10-1/2 inches deep. It weighs 16 pounds and has a 1 gallon steel tank with manual drain. The 14G824 can be used for inflation, stapling, brad nailing, air brushing and other light duty applications. This is an oil-free piston type compressor from Grainger.