Learn More
Grainger Panel Truck 3200 lb Cap 54inL 30inW
Catalog No. 502749346
Encompass_Preferred
The Grainger 14G822 is a heavy duty industrial grade bench vise that features a 6 inch jaw width capacity, 4 inch jaw opening capacity, and a 360 degree swivel base. It has an anvil working surface and is constructed from high strength cast iron with a powder coat finish. This vise can securely clamp a wide range of workpieces on a workbench or other sturdy surface. With its rugged design, the Grainger 14G822 bench vise is well suited for machine shops, garages, and industrial work settings.
Catalog No. 50-274-9346 Supplier Grainger Supplier No. 14G822
The Grainger 14G822 is a heavy duty industrial grade bench vise that features a 6 inch jaw width capacity, 4 inch jaw opening capacity, and a 360 degree swivel base. It has an anvil working surface and is constructed from high strength cast iron with a powder coat finish. This vise can securely clamp a wide range of workpieces on a workbench or other sturdy surface. With its rugged design, the Grainger 14G822 bench vise is well suited for machine shops, garages, and industrial work settings.