Learn More
Enzo Life Sciences Panepoxydone (0.5mg). CAS: 31298-54-1
SDP
Supplier:  Enzo Life Sciences ALX350109MC05
Encompass_Preferred
Antibiotic. Inhibitor of NF-κB activation by preventing the phosphorylation of IκB protein and thereby interrupts the signalling pathway. Strongly inhibits the expression of several NF-κB dependent pro-inflammatory genes. Purity: ≥95% (HPLC, MS). Appearance: Yellow oil. Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8736